Assay Details
| Gene Symbol: SPAG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285104.1 | NP_001272033.1 | 3 - 4 | 403 | 77 | |
| XM_005564788.2 | XP_005564845.1 | 3 - 4 | 411 | 77 | ||
| XM_005564790.2 | XP_005564847.1 | 3 - 4 | 471 | 77 | ||
| XM_005564791.2 | XP_005564848.1 | 3 - 4 | 471 | 77 | ||
| GenBank mRNA | AB169228.1 | - | 3 - 4 | 403 | 77 | |
| AB179314.1 | - | 3 - 4 | 403 | 77 | ||
More Information
Set Membership: |
|

SPAG6
Genomic Map