Assay Details
| Gene Symbol: GAPDHS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284052.1 | NP_001270981.1 | 10 - 11 | 1213 | 68 | |
| GenBank mRNA | AB179184.1 | - | 10 - 11 | 1213 | 68 | |
More Information
Set Membership: |
|

GAPDHS
Genomic Map