Assay Details
| Gene Symbol: IGSF21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441894.1 | XP_015297380.1 | 8 - 9 | 1256 | 56 | |
| XM_015441895.1 | XP_015297381.1 | 7 - 8 | 1175 | 56 | ||
| XM_015441901.1 | XP_015297387.1 | 8 - 9 | 1024 | 56 | ||
| GenBank mRNA | AB046009.1 | - | 8 - 9 | 1473 | 56 | |
More Information
Set Membership: |
|

IGSF21
Genomic Map