Assay Details
| Gene Symbol: G6PC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573395.1 | XP_005573452.1 | 1 - 2 | 346 | 97 | |
| XM_005573396.1 | XP_005573453.1 | 1 - 2 | 346 | 97 | ||
More Information
Set Membership: |
|

G6PC2
Genomic Map