Assay Details
| Gene Symbol: TCP11L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284946.1 | NP_001271875.1 | 5 - 6 | 869 | 96 | |
| XM_005578256.2 | XP_005578313.1 | 5 - 6 | 849 | 96 | ||
| XM_005578258.2 | XP_005578315.1 | 5 - 6 | 1235 | 96 | ||
| XM_015434086.1 | XP_015289572.1 | 5 - 6 | 914 | 96 | ||
| XM_015434087.1 | XP_015289573.1 | 4 - 5 | 685 | 96 | ||
| GenBank mRNA | AB171974.1 | - | 5 - 6 | 869 | 96 | |
More Information
Set Membership: |
|

TCP11L1
Genomic Map