Assay Details
| Gene Symbol: RAB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590943.2 | XP_005591000.1 | 6 - 7 | 684 | 62 | |
| XM_005590945.2 | XP_005591002.1 | 6 - 7 | 684 | 62 | ||
More Information
Set Membership: |
|

RAB26
Genomic Map