Assay Details
| Gene Symbol: ECPAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581082.2 | XP_005581139.1 | 28 - 29 | 3341 | 98 | |
| XM_005581083.2 | XP_005581140.1 | 27 - 28 | 3355 | 98 | ||
| GenBank mRNA | AB172431.1 | - | 10 - 11 | 1187 | 98 | |
More Information
Set Membership: |
|

ECPAS
Genomic Map