Assay Details
Gene Symbol: SYNGR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589759.2 | XP_005589816.1 | 4 - 5 | 665 | 69 | |
XM_005589760.2 | XP_005589817.1 | 4 - 5 | 756 | 69 | ||
XM_005589761.2 | XP_005589818.1 | 4 - 5 | 637 | 69 | ||
XM_005589762.2 | XP_005589819.1 | 4 - 5 | 620 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
