Assay Details
| Gene Symbol: LRRC46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284584.1 | NP_001271513.1 | 5 - 6 | 693 | 79 | |
| GenBank mRNA | AB168981.1 | - | 5 - 6 | 693 | 79 | |
More Information
Set Membership: |
|

LRRC46
Genomic Map