Assay Details
| Gene Symbol: TGFBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581203.2 | XP_005581260.1 | 3 - 4 | 1973 | 126 | |
| XM_015436934.1 | XP_015292420.1 | 3 - 4 | 1986 | 126 | ||
| XM_015436936.1 | XP_015292422.1 | 3 - 4 | 507 | 126 | ||
| XM_015436937.1 | XP_015292423.1 | 2 - 3 | 2539 | 126 | ||
| XM_015436938.1 | XP_015292424.1 | 3 - 4 | 541 | 126 | ||
More Information
Set Membership: |
|

TGFBR1
Genomic Map