Assay Details
| Gene Symbol: SMAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557844.2 | XP_005557901.1 | 2 - 3 | 693 | 150 | |
| XM_005557845.2 | XP_005557902.1 | 2 - 3 | 7471 | 150 | ||
| XM_005557846.2 | XP_005557903.1 | 2 - 3 | 693 | 150 | ||
| XM_005557848.2 | XP_005557905.1 | 2 - 3 | 1037 | 150 | ||
More Information
Set Membership: |
|

SMAD5
Genomic Map