Assay Details
| Gene Symbol: C1H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285128.1 | NP_001272057.1 | 2 - 3 | 357 | 99 | |
| XM_015452008.1 | XP_015307494.1 | 2 - 3 | 476 | 99 | ||
| GenBank mRNA | AB174639.1 | - | 2 - 3 | 357 | 99 | |

C1H1orf43
Genomic Map