Assay Details
| Gene Symbol: FDPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541536.2 | XP_005541593.1 | 2 - 3 | 342 | 91 | |
| XM_015456156.1 | XP_015311642.1 | 2 - 3 | 332 | 91 | ||
More Information
Set Membership: |
|

FDPS
Genomic Map