Assay Details
| Gene Symbol: TEAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553217.2 | XP_005553274.1 | 11 - 12 | 1209 | 58 | |
| XM_015449511.1 | XP_015304997.1 | 10 - 11 | 1146 | 58 | ||
| XM_015449512.1 | XP_015304998.1 | 10 - 11 | 1289 | 58 | ||
| XM_015449513.1 | XP_015304999.1 | 9 - 10 | 1057 | 58 | ||
| XM_015449514.1 | XP_015305000.1 | 10 - 11 | 1142 | 58 | ||
More Information
Set Membership: |
|

TEAD3
Genomic Map