Assay Details
| Gene Symbol: POFUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283140.1 | NP_001270069.1 | 3 - 4 | 483 | 78 | |
| XM_015457670.1 | XP_015313156.1 | 3 - 4 | 548 | 78 | ||
| GenBank mRNA | AB171323.1 | - | 3 - 4 | 483 | 78 | |
More Information
Set Membership: |
|

POFUT1
Genomic Map