Assay Details
| Gene Symbol: RO60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284903.1 | NP_001271832.1 | 5 - 6 | 1250 | 108 | |
| XM_005540276.2 | XP_005540333.1 | 6 - 7 | 1571 | 108 | ||
| XM_005540278.2 | XP_005540335.1 | 5 - 6 | 1198 | 108 | ||
| XM_005540279.2 | XP_005540336.1 | 5 - 6 | 1743 | 108 | ||
| XM_015445538.1 | XP_015301024.1 | 5 - 6 | 1687 | 108 | ||
| GenBank mRNA | AB174235.1 | - | 5 - 6 | 1250 | 108 | |
More Information
Set Membership: |
|

RO60
Genomic Map