Assay Details
| Gene Symbol: FAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555879.2 | XP_005555936.1 | 10 - 11 | 12897 | 103 | |
| XM_005555880.2 | XP_005555937.1 | 9 - 10 | 13322 | 103 | ||
| XM_015450808.1 | XP_015306294.1 | 8 - 9 | 13190 | 103 | ||
More Information
Set Membership: |
|

FAT4
Genomic Map