Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564559.2 | XP_005564616.1 | 12 - 13 | 1837 | 158 | |
| XM_005564565.2 | XP_005564622.1 | 12 - 13 | 1372 | 158 | ||
| XM_015455497.1 | XP_015310983.1 | 12 - 13 | 1228 | 158 | ||
| XM_015455498.1 | XP_015310984.1 | 11 - 12 | 1131 | 158 | ||
| XM_015455500.1 | XP_015310986.1 | 10 - 11 | 1035 | 158 | ||
| XM_015455501.1 | XP_015310987.1 | 10 - 11 | 993 | 158 | ||
| XM_015455502.1 | XP_015310988.1 | 11 - 12 | 1131 | 158 | ||
| XM_015455503.1 | XP_015310989.1 | 11 - 12 | 1454 | 158 | ||
| XM_015455505.1 | XP_015310991.1 | 11 - 12 | 1080 | 158 | ||
| XM_015455506.1 | XP_015310992.1 | 11 - 12 | 1141 | 158 | ||
| XM_015455507.1 | XP_015310993.1 | 10 - 11 | 1358 | 158 | ||
| XM_015455508.1 | XP_015310994.1 | 10 - 11 | 1316 | 158 | ||
| XM_015455509.1 | XP_015310995.1 | 9 - 10 | 1220 | 158 | ||
| XM_015455510.1 | XP_015310996.1 | 6 - 7 | 1245 | 158 | ||
More Information
Set Membership: |
|

TASOR2
Genomic Map