Assay Details
| Gene Symbol: C14H11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579533.2 | XP_005579590.1 | 5 - 6 | 301 | 138 | |
| XM_005579535.1 | XP_005579592.1 | 4 - 5 | 293 | 138 | ||
| XM_005579538.2 | XP_005579595.1 | 3 - 4 | 165 | 138 | ||
| XM_005579539.1 | XP_005579596.1 | 2 - 3 | 137 | 138 | ||
| XM_015435667.1 | XP_015291153.1 | 5 - 6 | 315 | 138 | ||
| XM_015435669.1 | XP_015291155.1 | 5 - 6 | 312 | 138 | ||
More Information
Set Membership: |
|

C14H11orf65
Genomic Map