Assay Details
| Gene Symbol: BCL2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560857.2 | XP_005560914.1 | 3 - 4 | 674 | 90 | |
| XM_005560858.2 | XP_005560915.1 | 3 - 4 | 736 | 90 | ||
More Information
Set Membership: |
|

BCL2L2
Genomic Map