Assay Details
Gene Symbol: GLRX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566755.2 | XP_005566812.1 | 7 - 8 | 813 | 151 | |
GenBank mRNA | AB174651.1 | - | 6 - 7 | 695 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
