Assay Details
Gene Symbol: LDB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566269.2 | XP_005566326.1 | 10 - 11 | 1117 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
