Assay Details
| Gene Symbol: NAA80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547199.2 | XP_005547256.1 | 1 - 2 | 218 | 140 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005547199.2 transcripts.
Set Membership: |
|

NAA80
Genomic Map