Assay Details
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571281.2 | XP_005571338.1 | 2 - 3 | 301 | 152 | |
| XM_015431073.1 | XP_015286559.1 | 2 - 3 | 301 | 152 | ||
More Information
Set Membership: |
|

NEMP1
Genomic Map