Assay Details
| Gene Symbol: PBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553407.2 | XP_005553464.1 | 4 | 864 | 113 | |
| XM_015449580.1 | XP_015305066.1 | 4 | 864 | 113 | ||
More Information
Set Membership: |
|

PBX2
Genomic Map