Assay Details
| Gene Symbol: GK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593230.2 | XP_005593287.1 | 1 - 2 | 199 | 151 | |
| XM_005593231.2 | XP_005593288.1 | 1 - 2 | 192 | 151 | ||
| XM_005593232.2 | XP_005593289.1 | 1 - 2 | 192 | 151 | ||
| XM_005593233.2 | XP_005593290.1 | 1 - 2 | 192 | 151 | ||
| XM_005593234.2 | XP_005593291.1 | 1 - 2 | 199 | 151 | ||
| XM_015443555.1 | XP_015299041.1 | 1 - 2 | 192 | 151 | ||
| XM_015443556.1 | XP_015299042.1 | 1 - 2 | 263 | 151 | ||
| XM_015443557.1 | XP_015299043.1 | 1 - 2 | 192 | 151 | ||
More Information
Set Membership: |
|

GK
Genomic Map