Assay Details
| Gene Symbol: STS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285408.1 | NP_001272337.1 | 4 - 5 | 824 | 52 | |
| GenBank mRNA | DQ266252.1 | - | 4 - 5 | 824 | 52 | |
| DQ529981.1 | - | 1 - 2 | 79 | 52 | ||
More Information
Set Membership: |
|

STS
Genomic Map