Assay Details
| Gene Symbol: CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449302.1 | XP_015304788.1 | 8 - 9 | 2912 | 71 | |
| XM_015449307.1 | XP_015304793.1 | 8 - 9 | 2912 | 71 | ||
| XM_015449309.1 | XP_015304795.1 | 7 - 8 | 2770 | 71 | ||
| XM_015449313.1 | XP_015304799.1 | 8 - 9 | 2179 | 71 | ||
| XM_015449318.1 | XP_015304804.1 | 8 - 9 | 2918 | 71 | ||
| XM_015449320.1 | XP_015304806.1 | 8 - 9 | 2180 | 71 | ||
| XM_015449323.1 | XP_015304809.1 | 8 - 9 | 2913 | 71 | ||
More Information
Set Membership: |
|

CR1
Genomic Map