Assay Details
| Gene Symbol: LGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565981.2 | XP_005566038.1 | 1 - 2 | 551 | 102 | |
| XM_005565982.2 | XP_005566039.1 | 1 - 2 | 551 | 102 | ||
| XM_015456401.1 | XP_015311887.1 | 1 - 2 | 551 | 102 | ||
More Information
Set Membership: |
|

LGI1
Genomic Map