Assay Details
| Gene Symbol: LOC102126451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582995.2 | XP_005583052.1 | 5 | 847 | 92 | |
| XM_005582997.2 | XP_005583054.1 | 4 | 708 | 92 | ||
| XM_015437704.1 | XP_015293190.1 | 4 | 2236 | 92 | ||
More Information
Set Membership: |
|

LOC102126451
Genomic Map