Assay Details
| Gene Symbol: CNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552387.2 | XP_005552444.1 | 2 | 1693 | 90 | |
| XM_005552388.2 | XP_005552445.1 | 3 | 1731 | 90 | ||
| XM_005552390.2 | XP_005552447.1 | 2 | 1539 | 90 | ||
More Information
Set Membership: |
|

CNR1
Genomic Map