Assay Details
| Gene Symbol: ATP5F1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569469.2 | XP_005569526.1 | 2 - 3 | 269 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP5F1E| Gene Symbol: ATP5F1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569469.2 | XP_005569526.1 | 2 - 3 | 269 | 90 | |
Set Membership: |
|