Assay Details
| Gene Symbol: LOC102144128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547321.2 | XP_005547378.1 | 2 | 6104 | 70 | |
| XM_015445980.1 | XP_015301466.1 | 9 | 3777 | 70 | ||
| GenBank mRNA | AB445678.1 | - | 1 | 2648 | 70 | |
More Information
Set Membership: |
|

LOC102144128
Genomic Map