Assay Details
| Gene Symbol: MICB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553568.2 | XP_005553625.1 | 2 - 3 | 693 | 120 | |
| XM_005553569.2 | XP_005553626.1 | 2 - 3 | 673 | 120 | ||
More Information
Set Membership: |
|

MICB
Genomic Map