Assay Details
| Gene Symbol: STX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549374.2 | XP_005549431.1 | 1 - 2 | 102 | 70 | |
| GenBank mRNA | AB170859.1 | - | 1 - 2 | 58 | 70 | |
More Information
Set Membership: |
|

STX1A
Genomic Map