Assay Details
| Gene Symbol: ICAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584689.1 | XP_005584746.1 | 4 - 5 | 924 | 71 | |
| XM_005584690.2 | XP_005584747.1 | 4 - 5 | 812 | 71 | ||
More Information
Set Membership: |
|

ICAM2
Genomic Map