Assay Details
| Gene Symbol: LOC102121984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587789.2 | XP_005587846.1 | 3 - 4 | 262 | 67 | |
| XM_015440843.1 | XP_015296329.1 | 3 - 4 | 325 | 67 | ||
| XM_015440844.1 | XP_015296330.1 | 4 - 5 | 453 | 67 | ||
| XM_015440845.1 | XP_015296331.1 | 3 - 4 | 260 | 67 | ||
More Information
Set Membership: |
|

LOC102121984
Genomic Map