Assay Details
| Gene Symbol: LOC102121984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587789.2 | XP_005587846.1 | 4 - 5 | 763 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102121984| Gene Symbol: LOC102121984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587789.2 | XP_005587846.1 | 4 - 5 | 763 | 120 | |
Set Membership: |
|