Assay Details
| Gene Symbol: CLEC7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570108.2 | XP_005570165.1 | 3 - 4 | 344 | 127 | |
| XM_015430367.1 | XP_015285853.1 | 3 - 4 | 347 | 127 | ||
| XM_015430369.1 | XP_015285855.1 | 3 - 4 | 346 | 127 | ||
More Information
Set Membership: |
|

CLEC7A
Genomic Map