Assay Details
| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285324.1 | NP_001272253.1 | 1 - 2 | 99 | 91 | |
| GenBank mRNA | AJ426430.1 | - | 1 - 2 | 99 | 91 | |
More Information
Set Membership: |
|

KLRF1
Genomic Map