Assay Details
| Gene Symbol: CD1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541336.2 | XP_005541393.1 | 5 - 6 | 1576 | 117 | |
| XM_015455159.1 | XP_015310645.1 | 4 - 5 | 2557 | 117 | ||
More Information
Set Membership: |
|

CD1E
Genomic Map