Assay Details
| Gene Symbol: CXCL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284543.1 | NP_001271472.1 | 2 - 3 | 239 | 65 | |
| GenBank mRNA | JQ288995.1 | - | 2 - 3 | 239 | 65 | |
More Information
Set Membership: |
|

CXCL13
Genomic Map