Assay Details
| Gene Symbol: IL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015451873.1 | XP_015307359.1 | 1 - 2 | 466 | 73 | |
| GenBank mRNA | AB000515.1 | - | 1 - 2 | 133 | 73 | |
More Information
Set Membership: |
|

IL4
Genomic Map