Assay Details
| Gene Symbol: REN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285029.1 | NP_001271958.1 | 2 - 3 | 286 | 63 | |
| GenBank mRNA | AY664490.1 | - | 2 - 3 | 303 | 63 | |
More Information
Set Membership: |
|

REN
Genomic Map