Assay Details
| Gene Symbol: CD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569956.2 | XP_005570013.1 | 4 - 5 | 621 | 64 | |
| XM_015430302.1 | XP_015285788.1 | 5 - 6 | 762 | 64 | ||
| GenBank mRNA | D63349.1 | - | 3 - 4 | 368 | 64 | |
More Information
Set Membership: |
|

CD4
Genomic Map