Assay Details
| Gene Symbol: KIF27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582030.2 | XP_005582087.1 | 16 - 17 | 3763 | 144 | |
| XM_005582034.2 | XP_005582091.1 | 16 - 17 | 3695 | 144 | ||
| XM_015437327.1 | XP_015292813.1 | 16 - 17 | 3808 | 144 | ||
| XM_015437328.1 | XP_015292814.1 | 16 - 17 | 3700 | 144 | ||
| GenBank mRNA | AB070138.1 | - | 12 - 13 | 1834 | 144 | |
More Information
Set Membership: |
|

KIF27
Genomic Map