Assay Details
| Gene Symbol: RCC1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549410.2 | XP_005549467.1 | 4 - 5 | 784 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RCC1L| Gene Symbol: RCC1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549410.2 | XP_005549467.1 | 4 - 5 | 784 | 68 | |
Set Membership: |
|