Assay Details
| Gene Symbol: MBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565842.2 | XP_005565899.1 | 3 - 4 | 449 | 69 | |
| XM_005565843.2 | XP_005565900.1 | 3 - 4 | 408 | 69 | ||
| XM_005565844.2 | XP_005565901.1 | 3 - 4 | 428 | 69 | ||
More Information
Set Membership: |
|

MBL2
Genomic Map