Assay Details
| Gene Symbol: SFTPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454563.1 | XP_015310049.1 | 4 - 5 | 496 | 91 | |
| XM_015454564.1 | XP_015310050.1 | 3 - 4 | 313 | 91 | ||
More Information
Set Membership: |
|

SFTPC
Genomic Map