Assay Details
| Gene Symbol: IL12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287275.1 | NP_001274204.1 | 1 - 2 | 107 | 65 | |
| XM_015451415.1 | XP_015306901.1 | 1 - 2 | 429 | 65 | ||
| GenBank mRNA | EF173869.1 | - | 1 - 2 | 52 | 65 | |
| JQ288992.1 | - | 1 - 2 | 107 | 65 | ||
More Information
Set Membership: |
|

IL12B
Genomic Map